COM448611371 Combi-Blocks, Inc.

tert-Butyl 4-[(3,4-difluorophenyl)methyl]piperazine-1-carboxylate

  • MDL number MFCD22383649
  • Cas Number : 1393441-76-3
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2162
Availability details for
Quantity
  • Availability
  • Add to Cart