COM448611373 Combi-Blocks, Inc.

Methyl 1-(2,2-dimethylpropanoyl)piperidine-4-carboxylate

  • MDL number MFCD03576207
  • Cas Number : 505088-49-3
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2165
Availability details for
Quantity
  • Availability
  • Add to Cart