COM448611387 Combi-Blocks, Inc.

2-Amino-N-tert-butyl-DL-propanamide

  • MDL number MFCD09936815
  • Cas Number : 83730-58-9
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2180
Availability details for
Quantity
  • Availability
  • Add to Cart