COM448611391 Combi-Blocks, Inc.

DL-2-Amino-1-(pyrrolidin-1-yl)propan-1-one

  • MDL number MFCD09929053
  • Cas Number : 56420-84-9
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2184
Availability details for
Quantity
  • Availability
  • Add to Cart