COM448611396 Combi-Blocks, Inc.

N-Propylpiperidine-4-carboxamide

  • MDL number MFCD08444678
  • Cas Number : 200267-73-8
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2190
Availability details for
Quantity
  • Availability
  • Add to Cart