COM448611404 Combi-Blocks, Inc.

N-tert-Butylpiperidine-4-carboxamide

  • MDL number MFCD06739717
  • Cas Number : 86542-86-1
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2198
Availability details for
Quantity
  • Availability
  • Add to Cart