COM448611408 Combi-Blocks, Inc.

t-Butyl N-[2-(2-chloro-2-phenylacetamido)ethyl]carbamate

  • MDL number MFCD22689077
  • Cas Number : 1420800-29-8
  • Molecular Weight : 312.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2205

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2205
Availability details for
Quantity
  • Availability
  • Add to Cart