COM448611427 Combi-Blocks, Inc.

tert-Butyl (3R)-3-ethoxypyrrolidine-1-carboxylate

  • MDL number MFCD08533290
  • Cas Number : 1212350-10-1
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2237
Availability details for
Quantity
  • Availability
  • Add to Cart