COM448611429 Combi-Blocks, Inc.

1,4-Bis(2-Aminophenyl)piperazine

  • MDL number MFCD18429864
  • Cas Number : 29549-88-0
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2240
Availability details for
Quantity
  • Availability
  • Add to Cart