COM448611439 Combi-Blocks, Inc.

tert-Butyl 4-[(pyridin-4-yl)carbonyl]piperidine-1-carboxylate

  • MDL number MFCD20261418
  • Cas Number : 1334415-27-8
  • Molecular Weight : 290.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2255
Availability details for
Quantity
  • Availability
  • Add to Cart