COM448611454 Combi-Blocks, Inc.

1-(Oxan-2-yl)pyrazole-5-methanamine

  • MDL number MFCD27935486
  • Cas Number : 1820642-31-6
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2290
Availability details for
Quantity
  • Availability
  • Add to Cart