COM448611469 Combi-Blocks, Inc.

N'-(1-Methylpiperidin-4-yl)(tert-butoxy)carbohydrazide

  • MDL number MFCD16496442
  • Cas Number : 1282030-14-1
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2313
Availability details for
Quantity
  • Availability
  • Add to Cart