COM448611483 Combi-Blocks, Inc.

1-(2-Nitrophenyl)piperidine-4-carboxamide

  • MDL number MFCD01604419
  • Cas Number : 923226-99-7
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2348
Availability details for
Quantity
  • Availability
  • Add to Cart