COM448611492 Combi-Blocks, Inc.

N-[(3R)-Piperidin-3-yl]benzamide

  • MDL number MFCD24553221
  • Cas Number : 1062136-14-4
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2388
Availability details for
Quantity
  • Availability
  • Add to Cart