COM448611500 Combi-Blocks, Inc.

3-Fluoro-4-piperazinobenzoic acid HCl

  • MDL number MFCD28385188
  • Cas Number : 1820639-99-3
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2402

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2402
Availability details for
Quantity
  • Availability
  • Add to Cart