COM448611503 Combi-Blocks, Inc.

1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD09260964
  • Cas Number : 111184-33-9
  • Molecular Weight : 128.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2410
Availability details for
Quantity
  • Availability
  • Add to Cart