COM448611513 Combi-Blocks, Inc.

4-Benzyloxy-2-fluoro-1-nitrobenzene

  • MDL number MFCD04115634
  • Cas Number : 221040-07-9
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1028
Availability details for
Quantity
  • Availability
  • Add to Cart