COM448611518 Combi-Blocks, Inc.

4-Benzyloxy-2-fluoroaniline, HCl

  • MDL number MFCD01632193
  • Cas Number : 478014-34-5
  • Molecular Weight : 253.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1033
Availability details for
Quantity
  • Availability
  • Add to Cart