COM448611522 Combi-Blocks, Inc.

2-Ethoxy-4-fluoroaniline, HCl

  • MDL number MFCD04115636
  • Cas Number : 850568-36-4
  • Molecular Weight : 191.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1037
Availability details for
Quantity
  • Availability
  • Add to Cart