COM448611525 Combi-Blocks, Inc.

3-Cyano-4-fluoroaniline, HCl

  • MDL number MFCD01632195
  • Cas Number : 801316-08-5
  • Molecular Weight : 172.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1041
Availability details for
Quantity
  • Availability
  • Add to Cart