COM448611541 Combi-Blocks, Inc.

4-Chloro-2-ethoxy-1-nitrobenzene

  • MDL number MFCD02625956
  • Cas Number : 29604-25-9
  • Molecular Weight : 201.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1057
Availability details for
Quantity
  • Availability
  • Add to Cart