COM448611550 Combi-Blocks, Inc.

t-Butyl 2-fluoro-4-iodophenyl(2-fluoro-4-iodophenylcarbamoyl)carbamate

  • MDL number MFCD10699602
  • Cas Number : 1033201-49-8
  • Molecular Weight : 600.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1068

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1068
Availability details for
Quantity
  • Availability
  • Add to Cart