COM448611553 Combi-Blocks, Inc.

4-Fluoro-3-methoxyaniline, HCl

  • MDL number MFCD08689481
  • Cas Number : 22510-10-7
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1071
Availability details for
Quantity
  • Availability
  • Add to Cart