COM448611575 Combi-Blocks, Inc.

4-Bromo-3-chloro-6-nitroanisole

  • MDL number MFCD09027196
  • Cas Number : 917562-21-1
  • Molecular Weight : 266.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1108
Availability details for
Quantity
  • Availability
  • Add to Cart