COM448611576 Combi-Blocks, Inc.

N-Formyl 2-bromo-4-fluoroaniline

  • MDL number MFCD08703393
  • Cas Number : 1495962-01-0
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1111
Availability details for
Quantity
  • Availability
  • Add to Cart