COM448611581 Combi-Blocks, Inc.

3-Benzylaniline, HCl

  • MDL number MFCD12756412
  • Cas Number : 6955-30-2
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1119
Availability details for
Quantity
  • Availability
  • Add to Cart