COM448611592 Combi-Blocks, Inc.

(N,N-Bis-t-Boc)-5-bromo-2-chloroaniline

  • MDL number MFCD09258633
  • Cas Number : 929000-06-6
  • Molecular Weight : 406.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1133
Availability details for
Quantity
  • Availability
  • Add to Cart