COM448611595 Combi-Blocks, Inc.

1-(2-Bromoethoxy)-2-nitrobenzene

  • MDL number MFCD02030589
  • Cas Number : 18800-37-8
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1136
Availability details for
Quantity
  • Availability
  • Add to Cart