COM448611608 Combi-Blocks, Inc.

2-Methylthio-5-nitrobenzonitrile

  • MDL number MFCD09475845
  • Cas Number : 74631-35-9
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1150
Availability details for
Quantity
  • Availability
  • Add to Cart