COM448611621 Combi-Blocks, Inc.

4-Chloro-2-methoxymethyl-N-methylaniline

  • MDL number MFCD09475903
  • Cas Number : 941294-12-8
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1169
Availability details for
Quantity
  • Availability
  • Add to Cart