COM448611628 Combi-Blocks, Inc.

4-Bromo-2,6-dichloro-3-methylaniline

  • MDL number MFCD09800852
  • Cas Number : 62406-68-2
  • Molecular Weight : 254.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1179
Availability details for
Quantity
  • Availability
  • Add to Cart