COM448611634 Combi-Blocks, Inc.

Methyl 1-(2-nitro-4-trifluoromethylphenyl)cyclopropanecarboxylate

  • MDL number MFCD09878310
  • Cas Number : 951885-67-9
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1189
Availability details for
Quantity
  • Availability
  • Add to Cart