COM448611650 Combi-Blocks, Inc.

N-(2-(4-Nitrophenoxy)ethyl)cyclopropanamine

  • MDL number MFCD10699603
  • Cas Number : 1033201-53-4
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1213
Availability details for
Quantity
  • Availability
  • Add to Cart