COM448611651 Combi-Blocks, Inc.

N,N-Dimethyl-2-(4-nitrophenoxy)ethanamine

  • MDL number MFCD01410114
  • Cas Number : 51344-13-9
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1214
Availability details for
Quantity
  • Availability
  • Add to Cart