COM448611653 Combi-Blocks, Inc.

N,N-Diethyl-2-(2-nitrophenoxy)ethanamine

  • MDL number MFCD01308707
  • Cas Number : 96115-77-4
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1220

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1220
Availability details for
Quantity
  • Availability
  • Add to Cart