COM448611660 Combi-Blocks, Inc.

3-Bromo-5-fluoro-4-methoxyaniline

  • MDL number MFCD08062384
  • Cas Number : 875664-44-1
  • Molecular Weight : 220.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1232
Availability details for
Quantity
  • Availability
  • Add to Cart