COM448611662 Combi-Blocks, Inc.

5-Acetamido-2-chlorophenol

  • MDL number MFCD09878312
  • Cas Number : 28443-52-9
  • Molecular Weight : 185.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1234

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1234
Availability details for
Quantity
  • Availability
  • Add to Cart