COM448611667 Combi-Blocks, Inc.

2,6-Dimethoxyaniline, HCl

  • MDL number MFCD09878313
  • Cas Number : 375397-36-7
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1245
Availability details for
Quantity
  • Availability
  • Add to Cart