COM448611683 Combi-Blocks, Inc.

N-Acetyl 3,5-dibromoaniline

  • MDL number MFCD09878317
  • Cas Number : 119430-40-9
  • Molecular Weight : 293.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1270
Availability details for
Quantity
  • Availability
  • Add to Cart