COM448611684 Combi-Blocks, Inc.

N-Acetyl 3,5-dibromo-2-nitroaniline

  • MDL number MFCD09972088
  • Cas Number : 855929-29-2
  • Molecular Weight : 338.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1271
Availability details for
Quantity
  • Availability
  • Add to Cart