COM448611686 Combi-Blocks, Inc.

Ethyl 2-(2-nitro-4-trifluoromethylphenylamino)acetate

  • MDL number MFCD00239175
  • Cas Number : 735-40-0
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1274
Availability details for
Quantity
  • Availability
  • Add to Cart