COM448611736 Combi-Blocks, Inc.

4-Chloro-3-ethoxyaniline

  • MDL number MFCD11053831
  • Cas Number : 852854-42-3
  • Molecular Weight : 171.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1353
Availability details for
Quantity
  • Availability
  • Add to Cart