COM448611737 Combi-Blocks, Inc.

2-(2,3-Difluoro-6-nitrophenyl)acetic acid

  • MDL number MFCD11053839
  • Cas Number : 141428-47-9
  • Molecular Weight : 217.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1354
Availability details for
Quantity
  • Availability
  • Add to Cart