COM448611740 Combi-Blocks, Inc.

3-Amino-2-hydroxymethylnaphthalene

  • MDL number MFCD00155332
  • Cas Number : 141281-58-5
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1359
Availability details for
Quantity
  • Availability
  • Add to Cart