COM448611743 Combi-Blocks, Inc.

3,4-Difluoro-2-methoxyaniline sulfate

  • MDL number MFCD11053865
  • Cas Number : 1072945-56-2
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1364
Availability details for
Quantity
  • Availability
  • Add to Cart