COM448611747 Combi-Blocks, Inc.

N-(3-Chlorophenyl) 3-aminobenzenesulfonamide

  • MDL number MFCD03147574
  • Cas Number : 438017-93-7
  • Molecular Weight : 282.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1368
Availability details for
Quantity
  • Availability
  • Add to Cart