COM448611758 Combi-Blocks, Inc.

2-Acetamido-1-nitro-3,5,6-trifluorobenzene

  • MDL number MFCD11504834
  • Cas Number : 388-11-4
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1384
Availability details for
Quantity
  • Availability
  • Add to Cart