COM448611760 Combi-Blocks, Inc.

2-Bromo-6-ethoxyaniline

  • MDL number MFCD11504835
  • Cas Number : 1072945-59-5
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1386
Availability details for
Quantity
  • Availability
  • Add to Cart