COM448611761 Combi-Blocks, Inc.

3-Amino-6-bromobiphenyl

  • MDL number MFCD11504836
  • Cas Number : 1036750-83-0
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1387
Availability details for
Quantity
  • Availability
  • Add to Cart