COM448611763 Combi-Blocks, Inc.

tert-Butyl 2-fluorophenylcarbamate

  • MDL number MFCD04112559
  • Cas Number : 98968-72-0
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-1389
Availability details for
Quantity
  • Availability
  • Add to Cart